Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNTNAP3
Entrez Gene ID
79937
Gene Name
contactin associated protein-like 3
Gene Aliases
CASPR3, CNTNAP3A
Chromosome Location
Chr.9: 39072767 - 39288303 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033655.3 NP_387504.2 21 - 22 3687 143
GenBank mRNA AF333769.2 - 21 - 22 3555 143
BC144130.1 - 19 - 20 3257 143

Gene Symbol
CNTNAP3B
Entrez Gene ID
728577
Gene Name
contactin associated protein-like 3B
Gene Aliases
-
Chromosome Location
Chr.9: 41891887 - 42129504 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201380.2 NP_001188309.2 21 - 22 3858 143

Gene Symbol
CNTNAP3P2
Entrez Gene ID
643827
Gene Name
contactin associated protein-like 3 pseudogene 2
Gene Aliases
-
Chromosome Location
Chr.9: 67059460 - 67297156 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_111893.2 21 - 22 3547 143

View Full Product Details