Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CNTNAP3
- Entrez Gene ID
-
79937
- Gene Name
- contactin associated protein-like 3
- Gene Aliases
-
CASPR3, CNTNAP3A
- Chromosome Location
-
Chr.9: 39072767 - 39288303 on Build GRCh38
- Gene Symbol
- CNTNAP3B
- Entrez Gene ID
-
728577
- Gene Name
- contactin associated protein-like 3B
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 41891887 - 42129504 on Build GRCh38
- Gene Symbol
- CNTNAP3P2
- Entrez Gene ID
-
643827
- Gene Name
- contactin associated protein-like 3 pseudogene 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 67059460 - 67297156 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_111893.2 |
|
9 - 10 |
1568 |
68 |
View Full Product Details