Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
PAGE5
Entrez Gene ID
90737
Gene Name
PAGE family member 5
Gene Aliases
CT16, CT16.1, CT16.2, GAGEE1, PAGE-5
Chromosome Location
Chr.X: 55220347 - 55224108 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013435.2 NP_001013453.1 1 - 1 56 133
GenBank mRNA BP429046.1 - 1 - 1 60 133
BQ437151.1 - 1 - 1 44 133

View Full Product Details