The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271969.1 | NP_001258898.1 | 2 - 3 | 395 | 122 |
NM_001271970.1 | NP_001258899.1 | 2 - 3 | 323 | 122 | |
NM_001271971.1 | NP_001258900.1 | 2 - 3 | 382 | 122 | |
NM_001271972.1 | NP_001258901.1 | 2 - 3 | 382 | 122 | |
NM_007355.3 | NP_031381.2 | 2 - 3 | 382 | 122 | |
GenBank mRNA | AK297367.1 | - | 2 - 3 | 239 | 122 |
AK312255.1 | - | 2 - 3 | 243 | 122 | |
AL136543.1 | - | 2 - 3 | 218 | 122 | |
AY359878.1 | - | 2 - 3 | 256 | 122 | |
BC004928.1 | - | 2 - 3 | 228 | 122 | |
BC007327.2 | - | 2 - 3 | 220 | 122 | |
BC012807.2 | - | 2 - 3 | 228 | 122 | |
BC014485.1 | - | 2 - 3 | 223 | 122 | |
BC016753.2 | - | 2 - 3 | 241 | 122 | |
BC068474.1 | - | 2 - 3 | 246 | 122 | |
BG482327.1 | - | 2 - 3 | 217 | 122 | |
BI252490.1 | - | 1 - 1 | 229 | 122 | |
BX420114.2 | - | 2 - 3 | 283 | 122 | |
BX649079.1 | - | 2 - 3 | 235 | 122 | |
CX761569.1 | - | 2 - 3 | 272 | 122 | |
DC341882.1 | - | 2 - 3 | 331 | 122 | |
DC350708.1 | - | 2 - 3 | 382 | 122 | |
DC365600.1 | - | 2 - 3 | 317 | 122 | |
M16660.1 | - | 2 - 3 | 228 | 122 |