Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM51
Entrez Gene ID
84767
Gene Name
tripartite motif-containing 51
Gene Aliases
SPRYD5, TRIM51A
Chromosome Location
Chr.11: 55883297 - 55891810 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032681.3 NP_116070.2 6 - 7 944 118
GenBank mRNA BC005014.1 - 4 - 5 584 118
JF968454.1 - 3 - 4 186 118

View Full Product Details