Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDC42
Entrez Gene ID
998
Gene Name
cell division cycle 42
Gene Aliases
CDC42Hs, G25K, TKS
Chromosome Location
Chr.1: 22052627 - 22092943 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039802.1 NP_001034891.1 6 - 7 777 94
NM_001791.3 NP_001782.1 5 - 6 651 94
GenBank mRNA AF498963.1 - 4 - 5 485 94
AK297465.1 - 5 - 6 734 94
AK303994.1 - 6 - 7 702 94
AK311375.1 - 4 - 5 495 94
AK316449.1 - 6 - 7 744 94
AL121735.1 - 5 - 6 589 94
BC002711.2 - 5 - 6 589 94
BC003682.1 - 4 - 5 538 94
BC018266.1 - 6 - 7 736 94
M57298.1 - 5 - 6 554 94

View Full Product Details