The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018654.1 | NP_061124.1 | 2 - 3 | 959 | 106 |
XM_005253421.3 | XP_005253478.1 | 4 - 5 | 3399 | 106 | |
XM_017019582.1 | XP_016875071.1 | 3 - 4 | 1492 | 106 | |
GenBank mRNA | AF209923.1 | - | 2 - 3 | 959 | 106 |
BC069341.1 | - | 2 - 3 | 959 | 106 | |
BC107077.1 | - | 2 - 3 | 959 | 106 | |
BC107078.1 | - | 2 - 3 | 959 | 106 |