Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
HBD
Entrez Gene ID
3045
Gene Name
hemoglobin subunit delta
Gene Aliases
-
Chromosome Location
Chr.11: 5232829 - 5234628 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000519.3 NP_000510.1 3 - 3 528 170
GenBank mRNA AY034468.1 - 3 - 3 528 170
BC069307.1 - 3 - 3 383 170
BC070282.1 - 3 - 3 384 170
BU664913.1 - 3 - 3 347 170

View Full Product Details