Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
INSL4
Entrez Gene ID
3641
Gene Name
insulin like 4
Gene Aliases
EPIL, PLACENTIN
Chromosome Location
Chr.9: 5231419 - 5233967 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002195.1 NP_002186.1 1 - 1 155 65
GenBank mRNA AK291543.1 - 1 - 1 155 65
BC026254.1 - 1 - 1 172 65
L34838.1 - 1 - 1 155 65

View Full Product Details