The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099682.2 | NP_001093152.2 | 4 - 5 | 496 | 108 |
XM_005278063.4 | XP_005278120.1 | 5 - 6 | 762 | 108 | |
XM_006724560.3 | XP_006724623.1 | 5 - 6 | 808 | 108 | |
GenBank mRNA | AI369342.1 | - | 1 - 2 | 108 | |
AK094251.1 | - | 3 - 4 | 1211 | 108 | |
AK096058.1 | - | 3 - 4 | 1819 | 108 | |
BC027995.1 | - | 4 - 5 | 465 | 108 | |
DQ884401.1 | - | 2 - 3 | 500 | 108 |