Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RRAS2
- Entrez Gene ID
-
22800
- Gene Name
- related RAS viral (r-ras) oncogene homolog 2
- Gene Aliases
-
TC21
- Chromosome Location
-
Chr.11: 14277920 - 14364506 on Build GRCh38
- Gene Symbol
- SCP2
- Entrez Gene ID
-
6342
- Gene Name
- sterol carrier protein 2
- Gene Aliases
-
NLTP, NSL-TP, SCP-2, SCP-CHI, SCP-X, SCPX
- Chromosome Location
-
Chr.1: 52927229 - 53051617 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BG028932.1 |
- |
- |
416 |
113 |
View Full Product Details