The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098529.1 | NP_001091999.1 | 1 - 1 | 346 | 142 |
GenBank mRNA | AK097656.1 | - | 1 - 1 | 96 | 142 |