Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SNORA47
- Entrez Gene ID
-
677828
- Gene Name
- small nucleolar RNA, H/ACA box 47
- Gene Aliases
-
HBI-115
- Chromosome Location
-
Chr.5: 77080434 - 77080571 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003014.2 |
|
1 - 1 |
71 |
83 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
zinc finger BED-type containing 3 |
ZBED3 |
|
|
Sense |
Chr. - 77076707 - 77087205 on Build GRCh38 |
Downstream |
ZBED3 antisense RNA 1 |
ZBED3-AS1 |
|
6.2kb
|
Anti-sense |
Chr. - 77086798 - 77148351 on Build GRCh38 |
View Full Product Details