Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SNORD69
- Entrez Gene ID
-
692109
- Gene Name
- small nucleolar RNA, C/D box 69
- Gene Aliases
-
HBII-210
- Chromosome Location
-
Chr.3: 52692736 - 52692812 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003057.1 |
|
1 - 1 |
38 |
69 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
small nucleolar RNA, C/D box 19B |
SNORD19B |
|
-1.9kb
|
Sense |
Chr. - 52690738 - 52690830 on Build GRCh38 |
Downstream |
glycosyltransferase 8 domain containing 1 |
GLT8D1 |
AD-017, MSTP139 |
1.7kb
|
Anti-sense |
Chr. - 52694484 - 52706083 on Build GRCh38 |
View Full Product Details