Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TSG1
- Entrez Gene ID
-
643432
- Gene Name
- tumor suppressor TSG1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.6: 93707083 - 93776481 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BX430875.2 |
- |
3 - 4 |
204 |
|
Chr. 6 - 93720245 - 93788632 [+] on Build GRCh38 |
96 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
EPH receptor A7 |
EPHA7 |
EHK-3, EHK3, EK11, HEK11 |
-317.0kb
|
Anti-sense |
Chr. - 93240020 - 93419687 on Build GRCh38 |
Downstream |
MANEA antisense RNA 1 (head to head) |
MANEA-AS1 |
|
1.8mb
|
Anti-sense |
Chr. - 95560096 - 95577451 on Build GRCh38 |
View Full Product Details