Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DYTN
Entrez Gene ID
391475
Gene Name
dystrotelin
Gene Aliases
-
Chromosome Location
Chr.2: 206651621 - 206718396 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001093730.1 NP_001087199.1 8 - 9 947 82
GenBank mRNA DQ516347.1 - 8 - 9 947 82

View Full Product Details