Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GGT6
Entrez Gene ID
124975
Gene Name
gamma-glutamyltransferase 6
Gene Aliases
-
Chromosome Location
Chr.17: 4556927 - 4560596 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122890.2 NP_001116362.1 1 - 2 217 116
NM_001288702.1 NP_001275631.1 1 - 2 217 116
NM_001288703.1 NP_001275632.1 1 - 2 217 116
NM_001288704.1 NP_001275633.1 1 - 2 217 116
NM_153338.3 NP_699169.2 1 - 2 217 116
GenBank mRNA AK074646.1 - 1 - 2 202 116
AK296645.1 - 1 - 2 198 116
AK300648.1 - 1 - 2 195 116
BC063111.1 - 1 - 2 187 116
BP276563.1 - 1 - 2 197 116
CV571037.1 - 1 - 2 165 116
DA645854.1 - 1 - 2 202 116
DC345075.1 - 1 - 2 217 116
DC345192.1 - 1 - 2 196 116

View Full Product Details