Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ZNF449
- Entrez Gene ID
-
203523
- Gene Name
- zinc finger protein 449
- Gene Aliases
-
ZSCAN19
- Chromosome Location
-
Chr.X: 135344098 - 135363413 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA336659.1 |
- |
1 - 2 |
|
|
Chr. X - 135344796 - 135347464 [+] on Build GRCh38 |
86 |
GenBankmRNA |
AY927532.3 |
- |
1 - 2 |
66 |
|
Chr. X - 135344541 - 135345868 [-] on Build GRCh38 |
86 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
cancer/testis antigen 55 |
CT55 |
BJHCC20A, CXorf48 |
-172.7kb
|
Sense |
Chr. - 135156536 - 135171827 on Build GRCh38 |
Downstream |
uncharacterized LOC100506790 |
LOC100506790 |
|
50.6kb
|
Anti-sense |
Chr. - 135396429 - 135397749 on Build GRCh38 |
View Full Product Details