Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRAM2
Entrez Gene ID
9697
Gene Name
translocation associated membrane protein 2
Gene Aliases
-
Chromosome Location
Chr.6: 52497402 - 52577064 on Build GRCh38
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY927571.1 - LIT3572
AY927571
2 - 2 451 133


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream EF-hand domain containing 1 EFHC1 EJM1, dJ304B14.2 -10.0kb Anti-sense Chr. - 52420196 - 52495785 on Build GRCh38
Downstream TRAM2 antisense RNA 1 (head to head) TRAM2-AS1 71.1kb Anti-sense Chr. - 52577202 - 52583993 on Build GRCh38

View Full Product Details