Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MIMT1
- Entrez Gene ID
-
100073347
- Gene Name
- MER1 repeat containing imprinted transcript 1 (non-protein coding)
- Gene Aliases
-
LINC00067, MIM1, NCRNA00067
- Chromosome Location
-
Chr.19: 56840902 - 56848554 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_024059.1 |
|
1 - 2 |
683 |
95 |
GenBank mRNA |
EF110915.1 |
- |
1 - 2 |
683 |
95 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
zinc finger imprinted 2 |
ZIM2 |
ZNF656 |
-173.0
|
Anti-sense |
Chr. - 56774547 - 56840729 on Build GRCh38 |
Downstream |
uncharacterized LOC105372474 |
LOC105372474 |
|
14.8kb
|
Sense |
Chr. - 56862413 - 56870571 on Build GRCh38 |
View Full Product Details