Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MKNK1
- Entrez Gene ID
-
8569
- Gene Name
- MAP kinase interacting serine/threonine kinase 1
- Gene Aliases
-
MNK1
- Chromosome Location
-
Chr.1: 46557407 - 46607140 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DC303144.1 |
- |
2 - 3 |
218 |
|
Chr. 1 - 46576625 - 46604279 [-] on Build GRCh38 |
104 |
GenBankmRNA |
DA764193.1 |
- |
2 - 3 |
219 |
|
Chr. 1 - 46576591 - 46604238 [-] on Build GRCh38 |
104 |
GenBankmRNA |
DC341091.1 |
- |
2 - 3 |
233 |
|
Chr. 1 - 46576596 - 46604252 [-] on Build GRCh38 |
104 |
GenBankmRNA |
BE908536.1 |
- |
1 - 2 |
148 |
|
Chr. 1 - 46580443 - 46594263 [-] on Build GRCh38 |
104 |
GenBankmRNA |
DA435703.1 |
- |
2 - 3 |
227 |
|
Chr. 1 - 46584547 - 46604246 [-] on Build GRCh38 |
104 |
GenBankmRNA |
DA470185.1 |
- |
2 - 3 |
188 |
|
Chr. 1 - 46576594 - 46604249 [-] on Build GRCh38 |
104 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
MKNK1 antisense RNA 1 |
MKNK1-AS1 |
|
-15.6kb
|
Anti-sense |
Chr. - 46538696 - 46570255 on Build GRCh38 |
Downstream |
MOB kinase activator 3C |
MOB3C |
MOB1E, MOBKL2C |
13.4kb
|
Sense |
Chr. - 46607715 - 46616891 on Build GRCh38 |
View Full Product Details