Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SIRT5
- Entrez Gene ID
-
23408
- Gene Name
- sirtuin 5
- Gene Aliases
-
SIR2L5
- Chromosome Location
-
Chr.6: 13574222 - 13615158 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC035122.1 |
- |
1 - 1 |
2354 |
77 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AI913731.1 |
- |
1 |
|
|
Chr. 6 - 13588333 - 13597058 [-] on Build GRCh38 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
glucose-fructose oxidoreductase domain containing 1 |
GFOD1 |
ADG-90, C6orf114 |
-107.0kb
|
Anti-sense |
Chr. - 13357830 - 13487637 on Build GRCh38 |
Downstream |
nucleolar protein 7 |
NOL7 |
C6orf90, PQBP3, RARG-1, dJ223E5.2 |
18.2kb
|
Sense |
Chr. - 13615327 - 13632470 on Build GRCh38 |
View Full Product Details