Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ICE1
- Entrez Gene ID
-
23379
- Gene Name
- interactor of little elongation complex ELL subunit 1
- Gene Aliases
-
KIAA0947
- Chromosome Location
-
Chr.5: 5422673 - 5490234 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC041002.2 |
- |
1 - 1 |
1742 |
104 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB326180.1 |
- |
1 |
|
|
Chr. 5 - 5488880 - 5489419 [-] on Build GRCh38 |
104 |
GenBankmRNA |
BM721129.1 |
- |
1 |
581 |
|
Chr. 5 - 5488402 - 5489184 [+] on Build GRCh38 |
104 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ADAM metallopeptidase with thrombospondin type 1 motif 16 |
ADAMTS16 |
ADAMTS16s |
-166.9kb
|
Sense |
Chr. - 5140330 - 5320304 on Build GRCh38 |
Downstream |
FLJ33360 protein |
FLJ33360 |
|
820.2kb
|
Anti-sense |
Chr. - 6310441 - 6337292 on Build GRCh38 |
View Full Product Details