Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RNPEP
- Entrez Gene ID
-
6051
- Gene Name
- arginyl aminopeptidase
- Gene Aliases
-
AP-B
- Chromosome Location
-
Chr.1: 201982638 - 202006147 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BX648806.1 |
- |
1 - 1 |
651 |
67 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CB990731.1 |
- |
|
|
|
Chr. - - [-] on Build |
67 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 6740 |
MIR6740 |
hsa-mir-6740 |
|
Anti-sense |
Chr. - 202003124 - 202003236 on Build GRCh38 |
Downstream |
E74 like ETS transcription factor 3 |
ELF3 |
EPR-1, ERT, ESE-1, ESX |
5.3kb
|
Anti-sense |
Chr. - 202010519 - 202017188 on Build GRCh38 |
View Full Product Details