Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TSL
- Entrez Gene ID
-
790953
- Gene Name
- testis-expressed, seven-twelve, leukemia
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 27615974 - 27627492 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_122122.1 |
|
2 - 3 |
1044 |
99 |
GenBank mRNA |
AB050001.1 |
- |
1 - 2 |
277 |
99 |
AB050003.1 |
- |
2 - 3 |
1043 |
99 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BX114521.1 |
- |
1 - 2 |
|
|
Chr. 7 - 27617498 - 27627492 [-] on Build GRCh38 |
99 |
GenBankmRNA |
AI581426.1 |
- |
1 - 2 |
|
|
Chr. 7 - 27617551 - 27627492 [-] on Build GRCh38 |
100 |
GenBankmRNA |
AI810001.1 |
- |
1 - 2 |
|
|
Chr. 7 - 27617509 - 27627492 [-] on Build GRCh38 |
99 |
GenBankmRNA |
AW104328.1 |
- |
1 - 2 |
|
|
Chr. 7 - 27617511 - 27627492 [-] on Build GRCh38 |
99 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
3-hydroxyisobutyrate dehydrogenase |
HIBADH |
NS5ATP1 |
|
Anti-sense |
Chr. - 27525440 - 27663001 on Build GRCh38 |
Downstream |
Tax1 binding protein 1 |
TAX1BP1 |
CALCOCO3, T6BP, TXBP151 |
111.9kb
|
Sense |
Chr. - 27739373 - 27829767 on Build GRCh38 |
View Full Product Details