The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_024393.1 | 1 - 2 | 503 | 65 | |
NR_117100.1 | 1 - 2 | 503 | 65 | ||
GenBank mRNA | DQ515896.1 | - | 1 - 2 | 503 | 65 |
DQ515897.1 | - | 1 - 2 | 503 | 65 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | POU class 5 homeobox 1B | POU5F1B | OCT4-PG1, OCT4PG1, OTF3C, OTF3P1, POU5F1P1, POU5F1P4, POU5FLC20, POU5FLC8 | -63.5kb | Anti-sense | Chr. - 127415612 - 127417210 on Build GRCh38 |
Downstream | cancer susceptibility candidate 11 (non-protein coding) | CASC11 | CARLo-7, LINC00990, TCONS_00014535 | 218.5kb | Sense | Chr. - 127700608 - 127733967 on Build GRCh38 |