Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CREM
- Entrez Gene ID
-
1390
- Gene Name
- cAMP responsive element modulator
- Gene Aliases
-
CREM-2, ICER, hCREM-2
- Chromosome Location
-
Chr.10: 35126841 - 35212958 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BU949828.1 |
- |
1 |
356 |
|
Chr. 10 - 35151968 - 35153727 [+] on Build GRCh38 |
76 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
cullin 2 |
CUL2 |
|
-25.4kb
|
Anti-sense |
Chr. - 35008551 - 35126899 on Build GRCh38 |
Downstream |
cyclin Y |
CCNY |
C10orf9, CBCP1, CCNX, CFP1 |
94.6kb
|
Sense |
Chr. - 35246981 - 35572670 on Build GRCh38 |
View Full Product Details