Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- RBM18
- Entrez Gene ID
-
92400
- Gene Name
- RNA binding motif protein 18
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 122239555 - 122264864 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027126.1 |
|
3 - 4 |
378 |
76 |
GenBank mRNA |
AK299625.1 |
- |
3 - 4 |
354 |
76 |
AK310807.1 |
- |
3 - 4 |
320 |
76 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DC414570.1 |
- |
3 - 4 |
320 |
|
Chr. 9 - 122245290 - 122264806 [-] on Build GRCh38 |
76 |
GenBankmRNA |
DC375338.1 |
- |
3 - 4 |
354 |
|
Chr. 9 - 122247518 - 122264839 [-] on Build GRCh38 |
76 |
GenBankmRNA |
BU171798.1 |
- |
3 - 4 |
308 |
|
Chr. 9 - 122241758 - 122264794 [-] on Build GRCh38 |
76 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
LIM homeobox 6 |
LHX6 |
LHX6.1 |
-23.0kb
|
Sense |
Chr. - 122185626 - 122228893 on Build GRCh38 |
Downstream |
mitochondrial ribosome recycling factor |
MRRF |
MRFF, MTRRF, RRF |
10.2kb
|
Anti-sense |
Chr. - 122264603 - 122323464 on Build GRCh38 |
View Full Product Details