Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TREML3P
Entrez Gene ID
340206
Gene Name
triggering receptor expressed on myeloid cells like 3, pseudogene
Gene Aliases
TLT3, TREML3
Chromosome Location
Chr.6: 41208554 - 41217947 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_027256.1 4 - 4 451 102
GenBank mRNA AF534825.1 - 4 - 4 451 102


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BG489188.1 - 340 Chr. - - [-] on Build 102


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream triggering receptor expressed on myeloid cells like 2 TREML2 C6orf76, TLT-2, TLT2, dJ238O23.1 -8.4kb Sense Chr. - 41189749 - 41201233 on Build GRCh38
Downstream triggering receptor expressed on myeloid cells like 4 TREML4 TLT-4, TLT4 18.6kb Anti-sense Chr. - 41228292 - 41239386 on Build GRCh38

View Full Product Details