The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025356.2 | NP_001020527.2 | 4 - 5 | 681 | 88 |
NM_001142678.1 | NP_001136150.1 | 4 - 5 | 528 | 88 | |
NM_001142679.1 | NP_001136151.1 | 4 - 5 | 681 | 88 | |
NM_001204803.1 | NP_001191732.1 | 5 - 6 | 744 | 88 | |
XM_005268706.4 | XP_005268763.1 | 4 - 5 | 635 | 88 | |
XM_005268707.3 | XP_005268764.1 | 3 - 4 | 555 | 88 | |
GenBank mRNA | AK126409.1 | - | 3 - 4 | 601 | 88 |
AK302819.1 | - | 5 - 6 | 538 | 88 | |
AK303996.1 | - | 4 - 5 | 612 | 88 | |
AK310641.1 | - | 4 - 5 | 648 | 88 | |
AL832340.1 | - | 4 - 5 | 528 | 88 | |
BC098410.1 | - | 4 - 5 | 548 | 88 | |
BC136445.1 | - | 5 - 6 | 437 | 88 |