Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANO6
Entrez Gene ID
196527
Gene Name
anoctamin 6
Gene Aliases
BDPLT7, SCTS, TMEM16F
Chromosome Location
Chr.12: 45215987 - 45440404 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025356.2 NP_001020527.2 7 - 8 1193 91
NM_001142678.1 NP_001136150.1 7 - 8 1040 91
NM_001142679.1 NP_001136151.1 7 - 8 1193 91
NM_001204803.1 NP_001191732.1 8 - 9 1256 91
XM_005268706.4 XP_005268763.1 7 - 8 1147 91
XM_005268707.3 XP_005268764.1 6 - 7 1067 91
GenBank mRNA AK126409.1 - 6 - 7 1113 91
AK302819.1 - 8 - 9 1050 91
AK303996.1 - 7 - 8 1124 91
AK310641.1 - 7 - 8 1160 91
AL832340.1 - 7 - 8 1040 91
BC098410.1 - 7 - 8 1060 91
BC136445.1 - 8 - 9 949 91

View Full Product Details