Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG205160.1 |
- |
1 |
|
|
Chr. 8 - 38772423 - 38772625 [-] on Build GRCh38 |
64 |
GenBankmRNA |
BG181899.1 |
- |
1 |
|
|
Chr. 8 - 38772378 - 38772642 [-] on Build GRCh38 |
64 |
GenBankmRNA |
BG210425.1 |
- |
1 |
|
|
Chr. 8 - 38772342 - 38772642 [-] on Build GRCh38 |
64 |
GenBankmRNA |
BG192866.1 |
- |
1 |
|
|
Chr. 8 - 38772326 - 38772642 [-] on Build GRCh38 |
64 |
GenBankmRNA |
BG197405.1 |
- |
1 |
|
|
Chr. 8 - 38772416 - 38772642 [-] on Build GRCh38 |
64 |
GenBankmRNA |
AL133564.1 |
- |
1 |
2451 |
|
Chr. 8 - 38770072 - 38772903 [+] on Build GRCh38 |
64 |
GenBankmRNA |
BG216119.1 |
- |
1 |
|
|
Chr. 8 - 38772419 - 38772642 [-] on Build GRCh38 |
64 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ring finger protein 5 pseudogene 1 |
RNF5P1 |
|
-168.8kb
|
Anti-sense |
Chr. - 38600173 - 38601257 on Build GRCh38 |
Downstream |
pleckstrin homology domain containing A2 |
PLEKHA2 |
TAPP2 |
128.3kb
|
Sense |
Chr. - 38901235 - 38973912 on Build GRCh38 |
View Full Product Details