Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SOCS2
- Entrez Gene ID
-
8835
- Gene Name
- suppressor of cytokine signaling 2
- Gene Aliases
-
CIS2, Cish2, SOCS-2, SSI-2, SSI2, STATI2
- Chromosome Location
-
Chr.12: 93569822 - 93626236 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AL161980.1 |
- |
1 - 1 |
4746 |
74 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BQ352731.1 |
- |
1 |
|
|
Chr. 12 - 93585281 - 93585491 [-] on Build GRCh38 |
74 |
GenBankmRNA |
N99179.1 |
- |
1 |
239 |
|
Chr. 12 - 93585144 - 93585678 [+] on Build GRCh38 |
74 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
SOCS2 antisense RNA 1 |
SOCS2-AS1 |
|
-9.2kb
|
Anti-sense |
Chr. - 93565628 - 93571398 on Build GRCh38 |
Downstream |
CASP2 and RIPK1 domain containing adaptor with death domain |
CRADD |
MRT34, RAIDD |
91.1kb
|
Sense |
Chr. - 93677342 - 93894840 on Build GRCh38 |
View Full Product Details