The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171653.1 | NP_001165124.1 | 9 - 9 | 8008 | 99 |
NM_014795.3 | NP_055610.1 | 10 - 10 | 8080 | 99 | |
XM_006712881.3 | XP_006712944.1 | 10 - 10 | 7674 | 99 | |
XM_006712882.3 | XP_006712945.1 | 10 - 10 | 7723 | 99 | |
XM_011512232.2 | XP_011510534.1 | 9 - 9 | 7717 | 99 | |
XM_017005415.1 | XP_016860904.1 | 9 - 9 | 8697 | 99 | |
GenBank mRNA | AF085983.1 | - | 1 - 1 | 453 | 99 |