Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DDIT3
- Entrez Gene ID
-
1649
- Gene Name
- DNA damage inducible transcript 3
- Gene Aliases
-
CEBPZ, CHOP, CHOP-10, CHOP10, GADD153
- Chromosome Location
-
Chr.12: 57516588 - 57520517 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC107859.1 |
- |
1 - 1 |
2148 |
82 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CR983205.1 |
- |
1 |
545 |
|
Chr. 12 - 57518003 - 57518912 [-] on Build GRCh38 |
82 |
GenBankmRNA |
DA647182.1 |
- |
1 |
284 |
|
Chr. 12 - 57517919 - 57518651 [-] on Build GRCh38 |
82 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 616 |
MIR616 |
MIRN616, hsa-mir-616 |
|
Sense |
Chr. - 57519163 - 57519259 on Build GRCh38 |
Downstream |
methyl-CpG binding domain protein 6 |
MBD6 |
|
195.0
|
Anti-sense |
Chr. - 57520710 - 57530148 on Build GRCh38 |
View Full Product Details