Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI125271.1 - 1 Chr. 10 - 46414155 - 46414613 [-] on Build GRCh38 92
GenBankmRNA BX492638.1 - 1 Chr. 10 - 46414330 - 46414874 [-] on Build GRCh38 92
GenBankmRNA AI306126.1 - 1 Chr. 10 - 47969955 - 47970326 [+] on Build GRCh38 92
GenBankmRNA DQ786233.1 - 1 1252 Chr. 10 - 47969950 - 47971373 [-] on Build GRCh38 92
GenBankmRNA CN264856.1 - 1 332 Chr. 10 - 47970019 - 47970453 [-] on Build GRCh38 92


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream annexin A8 ANXA8 ANX8, CH17-360D5.2 Sense Chr. - 47467993 - 47991879 on Build GRCh38
Downstream family with sequence similarity 25 member C FAM25C bA164N7.4 23.9kb Sense Chr. - 47995322 - 47999791 on Build GRCh38

View Full Product Details