Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CDKAL1
- Entrez Gene ID
-
54901
- Gene Name
- CDK5 regulatory subunit associated protein 1 like 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.6: 20534457 - 21232404 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK074342.1 |
- |
1 - 1 |
1254 |
80 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AW069867.1 |
- |
1 |
|
|
Chr. 6 - 21200158 - 21200588 [-] on Build GRCh38 |
80 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
E2F transcription factor 3 |
E2F3 |
E2F-3 |
-705.4kb
|
Sense |
Chr. - 20353285 - 20493714 on Build GRCh38 |
Downstream |
long intergenic non-protein coding RNA 581 |
LINC00581 |
linc-MBOAT1-4 |
285.5kb
|
Anti-sense |
Chr. - 21486061 - 21511892 on Build GRCh38 |
View Full Product Details