Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AA573118.1 |
- |
1 |
|
|
Chr. 8 - 94988568 - 94989166 [-] on Build GRCh38 |
93 |
GenBankmRNA |
BG740695.1 |
- |
1 |
|
|
Chr. 8 - 94988117 - 94988817 [+] on Build GRCh38 |
92 |
GenBankmRNA |
BM849607.1 |
- |
1 |
294 |
|
Chr. 8 - 94988352 - 94988796 [+] on Build GRCh38 |
93 |
GenBankmRNA |
AK074467.1 |
- |
1 |
1751 |
|
Chr. 8 - 94986881 - 94989189 [+] on Build GRCh38 |
93 |
GenBankmRNA |
CK902268.1 |
- |
2 |
339 |
|
Chr. 8 - 94980908 - 94988826 [+] on Build GRCh38 |
93 |
GenBankmRNA |
BQ777940.1 |
- |
2 |
340 |
|
Chr. 8 - 94980907 - 94988816 [+] on Build GRCh38 |
93 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
NADH:ubiquinone oxidoreductase complex assembly factor 6 |
NDUFAF6 |
C8orf38 |
|
Sense |
Chr. - 94895271 - 95118496 on Build GRCh38 |
Downstream |
uncharacterized LOC105375650 |
LOC105375650 |
|
75.0kb
|
Anti-sense |
Chr. - 95066808 - 95073182 on Build GRCh38 |
View Full Product Details