Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LSM8
- Entrez Gene ID
-
51691
- Gene Name
- LSM8 homolog, U6 small nuclear RNA associated
- Gene Aliases
-
NAA38
- Chromosome Location
-
Chr.7: 118184032 - 118204039 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK090561.1 |
- |
1 - 1 |
1252 |
96 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AL597401.1 |
- |
1 |
296 |
|
Chr. 7 - 118187763 - 118188294 [+] on Build GRCh38 |
96 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
cortactin binding protein 2 |
CTTNBP2 |
C7orf8, CORTBP2, Orf4 |
-313.3kb
|
Anti-sense |
Chr. - 117710648 - 117873507 on Build GRCh38 |
Downstream |
ankyrin repeat domain 7 |
ANKRD7 |
TSA806 |
35.4kb
|
Sense |
Chr. - 118224658 - 118242731 on Build GRCh38 |
View Full Product Details