Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BC013195.1 - 1 1751 Chr. 16 - 4335870 - 4337818 [-] on Build GRCh38 65


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream GLIS2 antisense RNA 1 GLIS2-AS1 -7.5kb Sense Chr. - 4324648 - 4328340 on Build GRCh38
Downstream presequence translocase-associated motor 16 homolog (S. cerevisiae) PAM16 CGI-136, MAGMAS, SMDMDM, TIM16, TIMM16 2.4kb Sense Chr. - 4340251 - 4351372 on Build GRCh38

View Full Product Details