Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SLC25A4
- Entrez Gene ID
-
291
- Gene Name
- solute carrier family 25 member 4
- Gene Aliases
-
AAC1, ANT, ANT 1, ANT1, MTDPS12, PEO2, PEO3, PEOA2, T1
- Chromosome Location
-
Chr.4: 185143263 - 185150384 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001151.3 |
NP_001142.2 |
4 - 4 |
4285 |
102 |
GenBank mRNA |
AF052119.1 |
- |
1 - 1 |
2249 |
102 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CB049452.1 |
- |
1 |
323 |
|
Chr. 4 - 185149940 - 185150384 [+] on Build GRCh38 |
102 |
GenBankmRNA |
BX117684.1 |
- |
1 |
311 |
|
Chr. 4 - 185149939 - 185150384 [+] on Build GRCh38 |
102 |
GenBankmRNA |
HY308996.1 |
- |
1 |
|
|
Chr. 4 - 185149890 - 185150382 [-] on Build GRCh38 |
102 |
GenBankmRNA |
AI955304.1 |
- |
1 |
|
|
Chr. 4 - 185149938 - 185150382 [-] on Build GRCh38 |
102 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC101928877 |
LOC101928877 |
|
-40.7kb
|
Sense |
Chr. - 185051876 - 185107239 on Build GRCh38 |
Downstream |
cilia and flagella associated protein 97 |
CFAP97 |
KIAA1430, hmw |
1.3kb
|
Anti-sense |
Chr. - 185159662 - 185210251 on Build GRCh38 |
View Full Product Details