Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SLC38A10
- Entrez Gene ID
-
124565
- Gene Name
- solute carrier family 38 member 10
- Gene Aliases
-
PP1744
- Chromosome Location
-
Chr.17: 81244811 - 81295433 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK098424.1 |
- |
1 - 1 |
645 |
72 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
HY377436.1 |
- |
1 |
|
|
Chr. 17 - 81251723 - 81252170 [+] on Build GRCh38 |
72 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
chromosome 17 open reading frame 89 |
C17orf89 |
|
-9.6kb
|
Anti-sense |
Chr. - 81239311 - 81241298 on Build GRCh38 |
Downstream |
long intergenic non-protein coding RNA 482 |
LINC00482 |
C17orf55 |
50.3kb
|
Sense |
Chr. - 81302824 - 81309248 on Build GRCh38 |
View Full Product Details