Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CB854393.1 - 1 Chr. 14 - 69231261 - 69231749 [-] on Build GRCh38 74
GenBankmRNA BX415971.2 - 1 Chr. 14 - 69230863 - 69231874 [-] on Build GRCh38 74
GenBankmRNA CR600957.1 - 1 1051 Chr. 14 - 69230544 - 69231874 [+] on Build GRCh38 74
GenBankmRNA CF886930.1 - 1 481 Chr. 14 - 69231114 - 69231829 [+] on Build GRCh38 74
GenBankmRNA CF890741.1 - 1 Chr. 14 - 69231260 - 69231927 [-] on Build GRCh38 74
GenBankmRNA AA527299.1 - 1 Chr. 14 - 69231397 - 69231912 [-] on Build GRCh38 74


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized LOC105370550 LOC105370550 Anti-sense Chr. - 69183018 - 69260567 on Build GRCh38
Downstream polypeptide N-acetylgalactosaminyltransferase 16 GALNT16 GALNACT16, GALNTL1, GalNAc-T16 27.7kb Sense Chr. - 69259546 - 69379904 on Build GRCh38

View Full Product Details