Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CR603341.1 - 1 153 Chr. 14 - 94635204 - 94635596 [+] on Build GRCh38 69


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream serpin family A member 3 SERPINA3 AACT, ACT, GIG24, GIG25 -11.2kb Sense Chr. - 94612377 - 94624053 on Build GRCh38
Downstream serpin family A member 13, pseudogene SERPINA13P SERPINA13, UNQ6121 5.1kb Sense Chr. - 94640725 - 94646994 on Build GRCh38

View Full Product Details