Assay Design

The assay probe spans an exon junction.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AL571349.3 - 4 - 5 Chr. 10 - 46759053 - 46786455 [+] on Build GRCh38 97
GenBankmRNA CR604707.1 - 1 - 2 1175 Chr. 10 - 46759053 - 46786811 [-] on Build GRCh38 96


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized LOC102724643 LOC102724643 -19.2kb Sense Chr. - 47587185 - 47619491 on Build GRCh38
Downstream FERM and PDZ domain containing 2B, pseudogene FRMPD2B FRMPD2L1, FRMPD2L2, FRMPD2P1, FRMPD2P2, FRMPDP2, PDZD5A, PDZD5B, PDZK5A, PDZK5B, bA556L1.2, yX59F3.2 89.9kb Anti-sense Chr. - 46876674 - 46895326 on Build GRCh38

View Full Product Details