Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CR605803.1 - 1 1350 Chr. 9 - 5429450 - 5431224 [-] on Build GRCh38 77
GenBankmRNA BX340463.2 - 1 Chr. 9 - 5429450 - 5430584 [+] on Build GRCh38 77


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream plasminogen receptor with a C-terminal lysine PLGRKT AD025, C9orf46, MDS030, PLG-RKT, Plg-R(KT) Sense Chr. - 5357966 - 5438476 on Build GRCh38
Downstream CD274 molecule CD274 B7-H, B7H1, PD-L1, PDCD1L1, PDCD1LG1, PDL1 19.2kb Anti-sense Chr. - 5450503 - 5470567 on Build GRCh38

View Full Product Details