Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DAPK2
Entrez Gene ID
23604
Gene Name
death associated protein kinase 2
Gene Aliases
DRP-1, DRP1
Chromosome Location
Chr.15: 63907036 - 64046470 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011521414.1 XP_011519716.1 11 - 11 2388 93
XM_017022044.1 XP_016877533.1 11 - 11 2384 93
GenBank mRNA AK094290.1 - 1 - 1 1446 93


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream microRNA 422a MIR422A MIRN422A -38.3kb Sense Chr. - 63870930 - 63871019 on Build GRCh38
Downstream uncharacterized LOC101928988 LOC101928988 17.0kb Anti-sense Chr. - 63928274 - 63935953 on Build GRCh38

View Full Product Details