Assay Design

The assay probe spans an exon junction.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK094916.1 - 1 - 2 69 Chr. 1 - 222724194 - 222773141 [-] on Build GRCh38 92
GenBankmRNA DA279987.1 - 1 - 2 69 Chr. 1 - 222729669 - 222773141 [-] on Build GRCh38 92


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream BRO1 domain and CAAX motif containing BROX C1orf58 Anti-sense Chr. - 222712553 - 222735196 on Build GRCh38
Downstream dispatched RND transporter family member 1 DISP1 DISPA 41.4kb Anti-sense Chr. - 222814514 - 223005995 on Build GRCh38

View Full Product Details