Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TSRM
- Entrez Gene ID
-
100287664
- Gene Name
- zinc finger domain-related protein TSRM
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY742823.1 |
- |
1 - 1 |
251 |
82 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
long intergenic non-protein coding RNA 998 |
LINC00998 |
|
-332.5kb
|
Anti-sense |
Chr. - 113116718 - 113118582 on Build GRCh38 |
Downstream |
protein phosphatase 1 regulatory subunit 3A |
PPP1R3A |
GM, PP1G, PPP1R3 |
425.4kb
|
Anti-sense |
Chr. - 113876827 - 113919094 on Build GRCh38 |
View Full Product Details