Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BE178284.1 |
- |
1 |
|
|
Chr. 2 - 37277278 - 37278028 [-] on Build GRCh38 |
99 |
GenBankmRNA |
BC043642.1 |
- |
1 |
2949 |
|
Chr. 2 - 37274911 - 37278239 [+] on Build GRCh38 |
99 |
GenBankmRNA |
BE177967.1 |
- |
1 |
587 |
|
Chr. 2 - 37277278 - 37277955 [+] on Build GRCh38 |
99 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
NADH:ubiquinone oxidoreductase complex assembly factor 7 |
NDUFAF7 |
C2orf56, MidA, PRO1853 |
-25.8kb
|
Sense |
Chr. - 37231631 - 37259052 on Build GRCh38 |
Downstream |
glutaminyl-peptide cyclotransferase |
QPCT |
GCT, QC, sQC |
66.4kb
|
Sense |
Chr. - 37344610 - 37373322 on Build GRCh38 |
View Full Product Details